Assay Design



Gene Transcripts

Gene Symbol
SGA2
Entrez Gene ID
842992
Gene Name
ASF1 like histone chaperone
Gene Aliases
ANTI- SILENCING FUNCTION 1A, ASF1A, AT1G66740, AtSP7, F4N21.13, F4N21_13, SP7
Chromosome Location
Chr.1: 24892340 - 24893876 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105344.4 NP_176846.1 2 - 3 472 67
GenBank mRNA AB078339.1 - 2 - 3 471 67
AY065298.1 - 2 - 3 470 67
AY096540.1 - 2 - 3 226 67

View Full Product Details