Assay Design



Gene Transcripts

Gene Symbol
ABA4
Entrez Gene ID
843028
Gene Name
abscisic acid (aba)-deficient 4
Gene Aliases
AT1G67080, F1O19.13, F1O19_13, abscisic acid (ABA)-deficient 4
Chromosome Location
Chr.1: 25045151 - 25047103 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001334271.1 NP_001321234.1 3 - 4 734 141
NM_105378.4 NP_564889.1 3 - 4 734 141
GenBank mRNA AK175554.1 - 3 - 4 360 141
AY084717.1 - 3 - 4 490 141

View Full Product Details