Assay Design
Gene Transcripts
- Gene Symbol
- ASY1
- Entrez Gene ID
-
843058
- Gene Name
- DNA-binding HORMA family protein
- Gene Aliases
-
ASYNAPTIC 1, AT1G67370, ATASY1, F1N21.19
- Chromosome Location
-
Chr.1: 25239117 - 25243837 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_105405.1 |
NP_564896.1 |
16 - 17 |
1308 |
100 |
View Full Product Details