Assay Design
Gene Transcripts
- Gene Symbol
- CLE12
- Entrez Gene ID
-
843211
- Gene Name
- CLAVATA3/ESR-RELATED 12
- Gene Aliases
-
AT1G68795, CLAVATA3/ESR-RELATED 12
- Chromosome Location
-
Chr.1: 25840626 - 25841686 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_105553.3 |
NP_564943.1 |
1 - 1 |
669 |
151 |
View Full Product Details