Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001334401.1 | NP_001320361.1 | 7 - 8 | 664 | 82 |
NM_105581.3 | NP_177074.1 | 7 - 8 | 910 | 82 | |
GenBank mRNA | BT004113.1 | - | 7 - 8 | 805 | 82 |
BT004951.1 | - | 7 - 8 | 663 | 82 |