Assay Design



Gene Transcripts

Gene Symbol
EXPA1
Entrez Gene ID
843288
Gene Name
expansin A1
Gene Aliases
AT-EXP1, AT1G69530, ATEXP1, ATEXPA1, ATHEXP ALPHA 1.2, EXP1, EXPANSIN 1, F10D13.18, F10D13_18, expansin A1
Chromosome Location
Chr.1: 26141726 - 26143538 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198428.1 NP_001185357.1 3 - 4 556 111
NM_001334412.1 NP_001319356.1 3 - 4 556 111
NM_105622.2 NP_177112.1 2 - 3 645 111
NM_179537.5 NP_849868.1 3 - 4 666 111
NM_179538.1 NP_849869.1 2 - 3 645 111
GenBank mRNA AF360291.1 - 3 - 4 554 111
AK318655.1 - 3 - 4 555 111
AY051047.1 - 2 - 3 447 111

View Full Product Details