Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001084331.3 | NP_001077800.1 | 8 - 9 | 1011 | 67 |
NM_001334421.1 | NP_001320950.1 | 7 - 8 | 1125 | 67 | |
NM_105642.4 | NP_177132.1 | 7 - 8 | 1037 | 67 | |
GenBank mRNA | AF327428.1 | - | 7 - 8 | 745 | 67 |
AF361803.1 | - | 7 - 8 | 781 | 67 | |
AY059154.1 | - | 6 - 7 | 716 | 67 | |
AY081254.1 | - | 7 - 8 | 787 | 67 | |
AY113941.1 | - | 6 - 7 | 716 | 67 | |
AY120705.1 | - | 7 - 8 | 745 | 67 | |
AY128711.1 | - | 6 - 7 | 716 | 67 |