Assay Design
Gene Transcripts
- Gene Symbol
- CMT3
- Entrez Gene ID
-
843313
- Gene Name
- chromomethylase 3
- Gene Aliases
-
AT1G69770, T6C23.3, T6C23_3, chromomethylase 3
- Chromosome Location
-
Chr.1: 26248118 - 26253687 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_105645.4 |
NP_177135.1 |
7 - 8 |
1054 |
109 |
View Full Product Details