Assay Design
Gene Transcripts
- Gene Symbol
- EDA24
- Entrez Gene ID
-
843391
- Gene Name
- Plant invertase/pectin methylesterase inhibitor superfamily protein
- Gene Aliases
-
AT1G70540, F24J13.11, F24J13_11, embryo sac development arrest 24
- Chromosome Location
-
Chr.1: 26593930 - 26594770 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_105722.3 |
NP_564990.1 |
1 - 1 |
551 |
152 |
View Full Product Details