Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001036185.1 | NP_001031262.1 | 6 - 7 | 616 | 86 |
NM_001036186.2 | NP_001031263.1 | 6 - 7 | 728 | 86 | |
NM_105726.3 | NP_177215.1 | 6 - 7 | 954 | 86 | |
NM_202393.2 | NP_974122.1 | 6 - 7 | 923 | 86 | |
GenBank mRNA | AF479640.1 | - | 5 - 6 | 488 | 86 |
AK316788.1 | - | 6 - 7 | 555 | 86 | |
AY035130.1 | - | 6 - 7 | 594 | 86 | |
AY062982.1 | - | 5 - 6 | 469 | 86 |