Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001334482.1 | NP_001321391.1 | 1 - 2 | 589 | 128 |
NM_105760.6 | NP_177248.3 | 2 - 3 | 501 | 128 | |
GenBank mRNA | AK118149.1 | - | 2 - 3 | 358 | 128 |
AK175837.1 | - | 2 - 3 | 358 | 128 | |
AK175965.1 | - | 2 - 3 | 358 | 128 | |
BT005594.1 | - | 2 - 3 | 269 | 128 | |
EU339186.1 | - | 2 - 3 | 439 | 128 |