Assay Design



Gene Transcripts

Gene Symbol
CSN5B
Entrez Gene ID
843463
Gene Name
COP9-signalosome 5B
Gene Aliases
AJH2, AT1G71230, COP9-signalosome 5B, CSN5, F3I17.12, F3I17_12
Chromosome Location
Chr.1: 26852406 - 26854419 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001334491.1 NP_001321294.1 5 - 6 932 73
NM_105792.2 NP_177279.1 4 - 5 1065 73
GenBank mRNA AF395061.1 - 4 - 5 911 73
AF411778.1 - 4 - 5 1010 73
AY124816.1 - 4 - 5 911 73

View Full Product Details