Assay Design



Gene Transcripts

Gene Symbol
PFI
Entrez Gene ID
843485
Gene Name
tubulin folding cofactor E / Pfifferling (PFI)
Gene Aliases
AT1G71440, F3I17.21, F3I17_21, PFIFFERLING, TFC E, TUBULIN-FOLDING COFACTOR E
Chromosome Location
Chr.1: 26921060 - 26924431 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105813.4 NP_565017.1 7 - 8 1038 70
GenBank mRNA AK118835.1 - 7 - 8 991 70
BT002403.1 - 7 - 8 993 70
BT008888.1 - 7 - 8 980 70

View Full Product Details