Assay Design



Gene Transcripts

Gene Symbol
AT1G71810
Entrez Gene ID
843511
Gene Name
Protein kinase superfamily protein
Gene Aliases
AT1G71810, F14O23.19, F14O23_19
Chromosome Location
Chr.1: 27002414 - 27008184 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105839.3 NP_565025.1 3 - 4 650 75
GenBank mRNA AY039896.1 - 3 - 4 553 75
AY094006.1 - 3 - 4 430 75

View Full Product Details