Assay Design



Gene Transcripts

Gene Symbol
SEC6
Entrez Gene ID
843512
Gene Name
SEC6
Gene Aliases
AT1G71820, F14O23.20, F14O23_20
Chromosome Location
Chr.1: 27009564 - 27017124 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198445.1 NP_001185374.1 18 - 19 2007 75
NM_105840.4 NP_565026.1 18 - 19 1926 75
GenBank mRNA AY046014.1 - 18 - 19 1847 75
AY133819.1 - 17 - 18 1578 75

View Full Product Details