Assay Design



Gene Transcripts

Gene Symbol
ERO1
Entrez Gene ID
843560
Gene Name
endoplasmic reticulum oxidoreductins 1
Gene Aliases
AERO1, AT1G72280, T9N14.18, T9N14_18, endoplasmic reticulum oxidoreductins 1
Chromosome Location
Chr.1: 27211821 - 27214688 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105887.4 NP_177372.1 1 - 2 352 86
GenBank mRNA AK227807.1 - 1 - 2 181 86
BT008774.1 - 1 - 2 170 86

View Full Product Details