Assay Design
Gene Transcripts
- Gene Symbol
- EXO70D1
- Entrez Gene ID
-
843579
- Gene Name
- exocyst subunit exo70 family protein D1
- Gene Aliases
-
AT1G72470, ATEXO70D1, T10D10.6, T10D10_6, exocyst subunit exo70 family protein D1
- Chromosome Location
-
Chr.1: 27283628 - 27285973 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_105906.4 |
NP_177391.1 |
1 - 1 |
2007 |
70 |
View Full Product Details