Assay Design



Gene Transcripts

Gene Symbol
CKL2
Entrez Gene ID
843603
Gene Name
casein kinase 1-like protein 2
Gene Aliases
AT1G72710, F28P22.10, F28P22_10, casein kinase 1-like protein 2
Chromosome Location
Chr.1: 27372197 - 27376730 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105930.3 NP_177415.1 12 - 13 1348 66
GenBank mRNA AY059850.1 - 12 - 13 1274 66
AY943843.1 - 12 - 13 992 66
BT010328.1 - 12 - 13 992 66

View Full Product Details