Assay Design



Gene Transcripts

Gene Symbol
CKL2
Entrez Gene ID
843603
Gene Name
casein kinase 1-like protein 2
Gene Aliases
AT1G72710, F28P22.10, F28P22_10, casein kinase 1-like protein 2
Chromosome Location
Chr.1: 27372197 - 27376730 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105930.3 NP_177415.1 8 - 9 986 80
GenBank mRNA AY059850.1 - 8 - 9 912 80
AY943843.1 - 8 - 9 630 80
BT010328.1 - 8 - 9 630 80

View Full Product Details