Assay Design
Gene Transcripts
- Gene Symbol
- scpl4
- Entrez Gene ID
-
843665
- Gene Name
- serine carboxypeptidase-like 4
- Gene Aliases
-
AT1G73310, T18K17.2, T18K17_2, serine carboxypeptidase-like 4
- Chromosome Location
-
Chr.1: 27562441 - 27565177 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_105990.2 |
NP_177474.1 |
12 - 13 |
1243 |
85 |
View Full Product Details