Assay Design
Gene Transcripts
- Gene Symbol
- CAT3
- Entrez Gene ID
-
833664
- Gene Name
- cationic amino acid transporter 3
- Gene Aliases
-
AT5G36940, MLF18.60, MLF18_60, cationic amino acid transporter 3
- Chromosome Location
-
Chr.5: 14590115 - 14594384 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_123052.5 |
NP_198510.2 |
13 - 14 |
1815 |
61 |
GenBank mRNA |
AK117747.1 |
- |
13 - 14 |
1620 |
61 |
View Full Product Details