Assay Design
Gene Transcripts
- Gene Symbol
- CHX24
- Entrez Gene ID
-
833677
- Gene Name
- cation/H+ exchanger 24
- Gene Aliases
-
ARABIDOPSIS THALIANA CATION/H+ EXCHANGER 24, AT5G37060, ATCHX24, MJG14.5, MJG14_5, cation/H+ exchanger 24
- Chromosome Location
-
Chr.5: 14642622 - 14645612 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_123065.2 |
NP_198522.1 |
1 - 2 |
494 |
69 |
View Full Product Details