Assay Design



Gene Transcripts

Gene Symbol
SEC
Entrez Gene ID
819579
Gene Name
Tetratricopeptide repeat (TPR)-like superfamily protein
Gene Aliases
AT3G04240, T6K12.14, T6K12_14, secret agent
Chromosome Location
Chr.3: 1113914 - 1120933 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111295.4 NP_187074.1 1 - 2 362 80
GenBank mRNA AF441079.1 - 1 - 2 280 80
AK226884.1 - 1 - 2 305 80
AY090938.1 - 1 - 2 301 80
AY117340.1 - 1 - 2 151 80
KJ138841.1 - 1 - 2 151 80

View Full Product Details