Assay Design



Gene Transcripts

Gene Symbol
SUS4
Entrez Gene ID
823393
Gene Name
sucrose synthase 4
Gene Aliases
ARABIDOPSIS THALIANA SUCROSE SYNTHASE 4, AT3G43190, ATSUS4, sucrose synthase 4
Chromosome Location
Chr.3: 15179010 - 15184105 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001339091.1 NP_001326640.1 10 - 11 2348 67
NM_114187.5 NP_566865.2 11 - 12 2530 67

View Full Product Details