Assay Design



Gene Transcripts

Gene Symbol
ADT4
Entrez Gene ID
823601
Gene Name
arogenate dehydratase 4
Gene Aliases
AT3G44720, arogenate dehydratase 4
Chromosome Location
Chr.3: 16271511 - 16273413 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_114340.4 NP_190058.1 1 - 1 1104 57
GenBank mRNA AY062692.1 - 1 - 1 963 57
BT008862.1 - 1 - 1 856 57
DQ411467.1 - 1 - 1 861 57

View Full Product Details