Assay Design
Gene Transcripts
- Gene Symbol
- CHX4
- Entrez Gene ID
-
823624
- Gene Name
- cation/H+ exchanger 4
- Gene Aliases
-
AT3G44900, ATCHX4, cation/H+ exchanger 4
- Chromosome Location
-
Chr.3: 16388724 - 16391582 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_114359.2 |
NP_190076.1 |
1 - 2 |
267 |
86 |
GenBank mRNA |
DQ499020.1 |
- |
1 - 2 |
267 |
86 |
View Full Product Details