Assay Design
Gene Transcripts
- Gene Symbol
- AIR3
- Entrez Gene ID
-
814953
- Gene Name
- Subtilisin-like serine endopeptidase family protein
- Gene Aliases
-
AT2G04160, AUXIN-INDUCED IN ROOT CULTURES 3
- Chromosome Location
-
Chr.2: 1401268 - 1407827 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_126453.5 |
NP_565309.2 |
6 - 7 |
1244 |
71 |
View Full Product Details