Assay Design



Gene Transcripts

Gene Symbol
ORG4
Entrez Gene ID
815155
Gene Name
OBP3-responsive protein 4 (ORG4)
Gene Aliases
AT2G06010, F5K7.23, F5K7_23, OBP3-responsive gene 4
Chromosome Location
Chr.2: 2336702 - 2338902 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_126615.5 NP_565333.1 1 - 2 274 63
GenBank mRNA AY072356.1 - 1 - 2 159 63
AY114726.1 - 1 - 2 135 63

View Full Product Details