Assay Design



Gene Transcripts

Gene Symbol
SCPL43
Entrez Gene ID
815721
Gene Name
serine carboxypeptidase-like 43
Gene Aliases
AT2G12480, T27D6.5, T27D6_5, serine carboxypeptidase-like 43
Chromosome Location
Chr.2: 5069544 - 5072656 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036274.1 NP_001031351.1 9 - 10 1171 111
NM_001335380.1 NP_001323740.1 9 - 10 1372 111
NM_001335381.1 NP_001323739.1 9 - 10 1300 111
NM_126893.4 NP_178937.2 9 - 10 1171 111
GenBank mRNA BT004306.1 - 9 - 10 1171 111

View Full Product Details