Assay Design



Gene Transcripts

Gene Symbol
ABH1
Entrez Gene ID
815840
Gene Name
ARM repeat superfamily protein
Gene Aliases
ABA HYPERSENSITIVE 1, AT2G13540, ATCBP80, CAP-BINDING PROTEIN 80, CBP80, ENS, ENSALADA, NUCLEAR CAP-BINDING PROTEIN CBP80, T10F5.8, T10F5_8
Chromosome Location
Chr.2: 5636884 - 5643066 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_126934.4 NP_565356.1 12 - 13 1308 89
GenBank mRNA AF268377.1 - 12 - 13 1308 89
AF272891.1 - 12 - 13 1175 89

View Full Product Details