Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001161044.1 | NP_001154516.1 | 5 - 6 | 944 | 130 |
NM_001335415.1 | NP_001323471.1 | 5 - 6 | 973 | 130 | |
NM_126984.4 | NP_565362.1 | 5 - 6 | 1099 | 130 | |
NM_126985.6 | NP_565363.2 | 5 - 6 | 944 | 130 |