Assay Design



Gene Transcripts

Gene Symbol
DRP3B
Entrez Gene ID
815898
Gene Name
dynamin related protein
Gene Aliases
ADL2B, AT2G14120, T22C12.5, T22C12_5, dynamin related protein
Chromosome Location
Chr.2: 5953940 - 5960263 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161044.1 NP_001154516.1 6 - 7 998 61
NM_001335415.1 NP_001323471.1 6 - 7 1027 61
NM_126984.4 NP_565362.1 6 - 7 1153 61
NM_126985.6 NP_565363.2 6 - 7 998 61

View Full Product Details