Assay Design



Gene Transcripts

Gene Symbol
AXR1
Entrez Gene ID
839286
Gene Name
NAD(P)-binding Rossmann-fold superfamily protein
Gene Aliases
AT1G05180, AUXIN RESISTANT 1, AUXIN RESISTANT PROTEIN 1, YUP8H12.21, YUP8H12_21
Chromosome Location
Chr.1: 1498114 - 1501953 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035893.1 NP_001030970.1 9 - 10 899 130
NM_100396.4 NP_172010.1 8 - 9 1126 130
NM_202032.2 NP_973761.1 9 - 10 899 130
GenBank mRNA AY050379.1 - 8 - 9 986 130
L13922.1 - 8 - 9 965 130

View Full Product Details