Assay Design



Gene Transcripts

Gene Symbol
APK1A
Entrez Gene ID
837271
Gene Name
Protein kinase superfamily protein
Gene Aliases
APK1, AT1G07570, F22G5.5, F22G5_5
Chromosome Location
Chr.1: 2331136 - 2333767 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001331708.1 NP_001318944.1 6 - 7 1138 72
NM_001331709.1 NP_001320609.1 5 - 6 1095 72
NM_001331710.1 NP_001320608.1 6 - 7 1044 72
NM_100631.4 NP_172237.1 5 - 6 1063 72
NM_202049.3 NP_973778.1 6 - 7 1092 72
GenBank mRNA BT004055.1 - 5 - 6 838 72
BT005112.1 - 5 - 6 758 72
D12522.1 - 5 - 6 835 72

View Full Product Details