Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_100960.3 | NP_563880.1 | 6 - 7 | 825 | 107 |
NM_202079.1 | NP_973808.1 | 5 - 6 | 711 | 107 | |
GenBank mRNA | AY054641.1 | - | 6 - 7 | 659 | 107 |
AY081546.1 | - | 5 - 6 | 508 | 107 | |
AY087338.1 | - | 6 - 7 | 727 | 107 | |
BT000765.1 | - | 6 - 7 | 604 | 107 |