Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_100960.3 | NP_563880.1 | 10 - 11 | 1122 | 62 |
NM_202079.1 | NP_973808.1 | 9 - 10 | 1008 | 62 | |
GenBank mRNA | AY054641.1 | - | 10 - 11 | 956 | 62 |
AY081546.1 | - | 9 - 10 | 805 | 62 | |
AY087338.1 | - | 10 - 11 | 1024 | 62 | |
BT000765.1 | - | 10 - 11 | 901 | 62 |