Assay Design



Gene Transcripts

Gene Symbol
SRS
Entrez Gene ID
837734
Gene Name
Seryl-tRNA synthetase
Gene Aliases
AT1G11870, ATSRS, F12F1.29, F12F1_29, OVA7, SERYL-TRNA SYNTHETASE, Seryl-tRNA synthetase, ovule abortion 7
Chromosome Location
Chr.1: 4003757 - 4007015 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332005.1 NP_001321932.1 4 - 5 695 106
NM_001332006.1 NP_001321931.1 4 - 5 695 106
NM_001332007.1 NP_001321930.1 2 - 3 305 106
NM_101058.2 NP_172651.1 4 - 5 640 106
NM_179316.3 NP_849647.1 4 - 5 735 106
NM_202083.1 NP_973812.1 4 - 5 646 106
GenBank mRNA AY091115.1 - 4 - 5 647 106
AY122978.1 - 4 - 5 597 106

View Full Product Details