Assay Design
Gene Transcripts
- Gene Symbol
- LTL1
- Entrez Gene ID
-
819584
- Gene Name
- Li-tolerant lipase 1
- Gene Aliases
-
AT3G04290, ATLTL1, Li-tolerant lipase 1, T6K12.9, T6K12_9
- Chromosome Location
-
Chr.3: 1133320 - 1136397 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_111300.4 |
NP_187079.1 |
1 - 2 |
413 |
79 |
GenBank mRNA |
AY085128.1 |
- |
1 - 2 |
306 |
79 |
View Full Product Details