Assay Design



Gene Transcripts

Gene Symbol
SUVR4
Entrez Gene ID
819593
Gene Name
SET-domain containing protein lysine methyltransferase family protein
Gene Aliases
AT3G04380, SDG31, SET DOMAIN PROTEIN 31, T27C4.2, T27C4_2
Chromosome Location
Chr.3: 1161366 - 1164977 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001337514.1 NP_001327806.1 6 - 7 1646 102
NM_111309.3 NP_187088.2 6 - 7 1633 102
NM_202488.2 NP_974217.1 7 - 8 1552 102

View Full Product Details