Assay Design



Gene Transcripts

Gene Symbol
MIRO3
Entrez Gene ID
819695
Gene Name
MIRO-related GTP-ase 3
Gene Aliases
AT3G05310, MIRO-related GTP-ase 3, T12H1.28, T12H1_28
Chromosome Location
Chr.3: 1510160 - 1513301 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001337576.1 NP_001327040.1 8 - 9 1644 81
NM_111404.1 NP_187182.1 10 - 11 1880 81

View Full Product Details