Assay Design
Gene Transcripts
- Gene Symbol
- MIRO3
- Entrez Gene ID
-
819695
- Gene Name
- MIRO-related GTP-ase 3
- Gene Aliases
-
AT3G05310, MIRO-related GTP-ase 3, T12H1.28, T12H1_28
- Chromosome Location
-
Chr.3: 1510160 - 1513301 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_111404.1 |
NP_187182.1 |
2 - 3 |
346 |
133 |
View Full Product Details