Assay Design



Gene Transcripts

Gene Symbol
MIRO3
Entrez Gene ID
819695
Gene Name
MIRO-related GTP-ase 3
Gene Aliases
AT3G05310, MIRO-related GTP-ase 3, T12H1.28, T12H1_28
Chromosome Location
Chr.3: 1510160 - 1513301 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001337575.1 NP_001327041.1 1 - 2 251 98
NM_001337576.1 NP_001327040.1 1 - 2 251 98
NM_111404.1 NP_187182.1 3 - 4 487 98

View Full Product Details