Assay Design



Gene Transcripts

Gene Symbol
ATN
Entrez Gene ID
819697
Gene Name
cyclin family
Gene Aliases
A. THALIANA TAN1, AT3G05330, ATTAN, T12H1.30, T12H1_30, TANGLED
Chromosome Location
Chr.3: 1518857 - 1521909 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111406.5 NP_187184.2 1 - 2 831 76
GenBank mRNA AK175941.1 - 1 - 2 416 76
AK227452.1 - 1 - 2 377 76
BT006478.1 - 1 - 2 81 76

View Full Product Details