Assay Design



Gene Transcripts

Gene Symbol
RAD9
Entrez Gene ID
819713
Gene Name
cell cycle checkpoint control protein family
Gene Aliases
AT3G05480, ATRAD9, F22F7.7, F22F7_7
Chromosome Location
Chr.3: 1585272 - 1589292 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001337585.1 NP_001319478.1 3 - 4 447 99
NM_001337586.1 NP_001319479.1 3 - 4 430 99
GenBank mRNA BT029213.1 - 3 - 4 336 99

View Full Product Details