Assay Design



Gene Transcripts

Gene Symbol
CHR11
Entrez Gene ID
819814
Gene Name
chromatin-remodeling protein 11
Gene Aliases
AT3G06400, F24P17.13, F24P17_13, chromatin-remodeling protein 11
Chromosome Location
Chr.3: 1940907 - 1946980 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202897.1 NP_001189826.1 18 - 19 2499 134
NM_111515.5 NP_187291.2 18 - 19 2559 134

View Full Product Details