Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001125119.2 | NP_001118591.1 | 6 - 7 | 1599 | 88 |
NM_111527.3 | NP_566285.1 | 7 - 8 | 1580 | 88 | |
GenBank mRNA | AJ491320.1 | - | 7 - 8 | 1496 | 88 |
AY059889.1 | - | 7 - 8 | 1421 | 88 | |
BT002100.1 | - | 7 - 8 | 1376 | 88 |