Assay Design



Gene Transcripts

Gene Symbol
IBM1
Entrez Gene ID
819952
Gene Name
Transcription factor jumonji (jmjC) domain-containing protein
Gene Aliases
AT3G07610, increase in bonsai methylation 1
Chromosome Location
Chr.3: 2425884 - 2433113 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084652.2 NP_001078121.1 1 - 2 719 79
NM_001202909.1 NP_001189838.1 1 - 2 534 79
NM_111640.2 NP_187418.1 1 - 2 719 79

View Full Product Details