Assay Design



Gene Transcripts

Gene Symbol
GRF8
Entrez Gene ID
828515
Gene Name
growth-regulating factor 8
Gene Aliases
AT4G24150, AtGRF8, AtMIN10, T19F6.140, T19F6_140, growth-regulating factor 8
Chromosome Location
Chr.4: 12535972 - 12539576 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001341644.1 NP_001329478.1 1 - 2 71 99
NM_118547.2 NP_194146.1 1 - 2 71 99

View Full Product Details