Assay Design



Gene Transcripts

Gene Symbol
cpHsc70-1
Entrez Gene ID
828531
Gene Name
chloroplast heat shock protein 70-1
Gene Aliases
AT4G24280, T22A6.110, T22A6_110, chloroplast heat shock protein 70-1
Chromosome Location
Chr.4: 12589810 - 12593800 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_118561.3 NP_194159.1 1 - 2 864 111
GenBank mRNA AY072138.1 - 1 - 2 687 111
BT001950.1 - 1 - 2 580 111

View Full Product Details