Assay Design



Gene Transcripts

Gene Symbol
PWD
Entrez Gene ID
828547
Gene Name
phosphoglucan, water dikinase
Gene Aliases
AT4G24450, ATGWD2, GWD3, T22A6.280, T22A6_280, phosphoglucan, water dikinase
Chromosome Location
Chr.4: 12635061 - 12642808 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001341673.1 NP_001328878.1 12 - 13 1606 66
NM_001341674.1 NP_001328879.1 12 - 13 1610 66
NM_118578.5 NP_194176.3 11 - 12 1408 66

View Full Product Details